Learn More
Memmert™ Natural Convection Hot Air Sterilizer with SingleDISPLAY
Shop All Memmert Products
Click to view available options
Capacity (Metric):
32 L
53 L
74 L
108 L
161 L
256 L
449 L
749 L
Unit Size:
Each
Description
- German, English, Spanish, French, Polish, Czech, Hungarian language settings
- SingleDISPLAY provides adaptive multifunctional digital PID-microprocessor controller with high-definition TFT-color display
- Digital backwards counter with target time setting, adjustable from 1 minute to 99 days
- Process time does not start until the set temperature is reached
- Three freely selectable temperature values
- Adjust temperature (Celsius or Fahrenheit), air flap position, program time, time zones, summertime/wintertime
Specifications
Specifications
| Certifications/Compliance | CE, cUL, DVE, EAC |
| Capacity (Metric) | 53 L |
| Type | Natural Convection Hot Air Sterilizer |
| Display | TFT |
| For Use With (Application) | Applied for sterilisation of medical materials, complete killing of highly resistant microorganisms, special application of depyrogenisation |
| Includes | Works Calibration Certificate |
| Material | Stainless Steel |
| Model | SN55 |
| Dimensions (L x W x H) | 585 x 784 x 514 mm |
| Length (Metric) | 514 mm |
| Show More |