Learn More
Thermo Scientific™ National Screw Thread Caps with Bonded Septa
Click to view available options
Pre-slit:
No
Yes
Closure Size:
10-425
13-425
8-425
Unit Size:
Pack of 100
Description
Effectively eliminate septa push-through with Thermo Scientific™ National Screw Thread Caps with Bonded Septa.
- Septa and polypropylene caps bonded together for an integral system
- Eliminates septa push-through
- Available with choice of GPI thread sizes and preslit septa for easier thin-gauge needle penetration
Specifications
Specifications
| Description | Black; Bonded Preslit Red 0.005 in. PTFE/White Silicone Septa; GPI Thread Finish: 10-425 |
| Color | White, Black |
| Material | PTFE, Silicone, Polypropylene |
| Pre-slit | Yes |
| Septum | Bonded Red PTFE/White Silicone, 0.005 in. |
| Product Line | National |
| Closure Size | 10-425 |