Learn More
Kautex™ Narrow Neck LDPE Bottles, Natural
Click to view available options
Capacity (Metric):
3 mL
10 mL
20 mL
50 mL
100 mL
200 mL
250 mL
1000 mL
Unit Size:
Each
Pack of 1500
Pack of 1800
Pack of 3600
Description
- Without closure
- Narrow mouth
- LDPE construction
- Metric capacity
Specifications
Specifications
| Capacity (Metric) | 10 mL |
| Material | LDPE |
| No. per Case | 1 Ea. |
| Packaging | Bottle |
| Quantity | 1 Ea. |
| Mouth | Narrow |
| Height (Metric) | 46.5 mm |
| Product Type | Narrow Mouth Bottle |
| Diameter (Metric) | 26.0 mm |