missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Benzyloxycarbonyl-L-2,3-diaminopropionic acid, 98%, Thermo Scientific Chemicals

Product Code. 15456936
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Nalpha-Benzyloxycarbonyl-L-2,3-diaminopropionic acid is a important building block for the synthesis of peptides containing DAP residues, e.g. bleomycins, edeines, tuberactinomycins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nalpha-Benzyloxycarbonyl-L-2,3-diaminopropionic acid is a important building block for the synthesis of peptides containing DAP residues, e.g. bleomycins, edeines, tuberactinomycins.

Solubility
Soluble in dilute acid.

Notes
Hygroscopic. Store in cool, well ventilated area. Keep container tightly closed. Store away from water/moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35761-26-3
Molecular Formula C11H14N2O4
Molecular Weight (g/mol) 238.243
MDL Number MFCD00237345
InChI Key FOXRXVSTFGNURG-VIFPVBQESA-N
Synonym z-dap-oh, cbz-beta-amino-l-alanine, 3-amino-2-n-cbz-l-alanine, s-3-amino-2-benzyloxy carbonyl amino propanoic acid, cbz-l-dap-oh, s-3-amino-2-benzyloxycarbonylamino propanoic acid, cbz-dap-oh, h-dap z-oh, n-alpha-cbz-l-2,3-diaminopropionic acid, z-dpr-oh
PubChem CID 2756313
IUPAC Name (2S)-3-amino-2-(phenylmethoxycarbonylamino)propanoic acid
SMILES C1=CC=C(C=C1)COC(=O)NC(CN)C(=O)O

Specifications

Melting Point ∼240°C (decomposition)
Beilstein 4486950
Solubility Information Soluble in dilute acid.
Quantity 1 g
Formula Weight 238.24
Optical Rotation −39° (c-1 in 1M HCl)
Sensitivity Hygroscopic
Percent Purity 98%
Chemical Name or Material Nalpha-Benzyloxycarbonyl-L-2,3-diaminopropionic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.