Learn More
Thermo Scientific™ Nalgene™ Polypropylene Pinch Clamp
From completely open to restricted flow to completely closed—experience precise, one-hand regulation of fluid flow with the Thermo Scientific™ Nalgene™ Polypropylene Pinch Clamp.
Brand: Thermo Scientific™ 6165-0002
104.53 EUR valid until 2025-06-13
Use promo code "24123" to get your promotional price.
Description
From completely open to restricted flow to completely closed—experience precise, one-hand regulation of fluid flow with the Thermo Scientific™ Nalgene™ Polypropylene Pinch Clamp. This plastic clamp will not rust, corrode or deform. Molded in one piece with no sharp edges, it accommodates a wide range of tubing.
Specifications
Case of 72 | |
Polypropylene | |
Pinch clamp; Fits tubing with O.D.: 1/4 to 7/16 in. |
Pinch Clamp | |
Fits tubing with O.D.: 1/4 to 7/16 in. |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
7448154010 | Certificate of Compliance | 10/04/2025 | 6165-0002 |
7452530010 | Certificate of Compliance | 31/03/2025 | 6165-0002 |
7423543010 | Certificate of Compliance | 17/03/2025 | 6165-0002 |
7353857010 | Certificate of Compliance | 17/03/2025 | 6165-0002 |
7415295010 | Certificate of Compliance | 17/03/2025 | 6165-0002 |