Learn More
Aalborg™ N2 Mass Flowmeter
Brand:  Aalborg™ GFM37A-VAL6-A0/3NC-33-SS
 View more versions of this product
Product Code. 11892370
  • € 1305.00 / Each
Estimated Shipment: 19-03-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Mass Flowmeter
0 to 50 LPM
±1.5 %
5°C to 50°C
±5%
12 V to 200 mA max
Full scale
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Special Offers
Special Offers