missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Diisopropylethylenediamine, 97%
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Applications
N,N-Diisopropylethylenediamine can be used to produce 1,4-diisopropyl-piperazin-2-one by heating. It is used as ligands for metathesis catalysts. It acts as anti-microbial agent.
Solubility
Fully miscible in water.
Notes
Air Sensitive. Store away from air, oxidizing agents. Incompatible materials are Strong oxidizing agents, Strong acids
N,N-Diisopropylethylenediamine can be used to produce 1,4-diisopropyl-piperazin-2-one by heating. It is used as ligands for metathesis catalysts. It acts as anti-microbial agent.
Solubility
Fully miscible in water.
Notes
Air Sensitive. Store away from air, oxidizing agents. Incompatible materials are Strong oxidizing agents, Strong acids
Specifications
Specifications
| Density | 0.828 |
| Boiling Point | 170°C |
| Flash Point | 54°C (129°F) |
| Refractive Index | 1.442 |
| Quantity | 10 g |
| UN Number | UN2733 |
| Beilstein | 1734800 |
| Sensitivity | Air Sensitive |
| Solubility Information | Fully miscible in water. |
| Formula Weight | 144.26 |
| Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction