missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-(2-morpholinopyrimidin-5-yl)methylamine, 97%, Thermo Scientific™

Product Code. p-3586856
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 10113224

Brand: Thermo Scientific Maybridge CC69546CB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 937796-14-0
Molecular Formula C10H16N4O
Molecular Weight (g/mol) 208.265
MDL Number MFCD09966161
InChI Key PFYPMSRZQMFCJO-UHFFFAOYSA-N
Synonym n-methyl-2-morpholinopyrimidin-5-yl methylamine, methyl 2-morpholin-4-yl pyrimidin-5-yl methyl amine, methyl 2-morpholin-4-ylpyrimidin-5-yl methyl amine, n-methyl-1-2-morpholin-4-yl pyrimidin-5-yl methanamine
PubChem CID 42556099
IUPAC Name N-methyl-1-(2-morpholin-4-ylpyrimidin-5-yl)methanamine
SMILES CNCC1=CN=C(N=C1)N2CCOCC2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 208.26
Percent Purity 97%
Chemical Name or Material N-methyl-(2-morpholinopyrimidin-5-yl)methylamine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.