missing translation for 'onlineSavingsMsg'
Learn More
n-Hydroxy-butyramidine, ≥97%, Thermo Scientific™ Product Code.: 10533423

n-Hydroxy-butyramidine, ≥97%, Thermo Scientific™

Product Code. 10533423
10 g, Amber Glass Bottle
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10533423

Brand: Thermo Scientific Maybridge MO07267EA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 27620-10-6
Molecular Formula C4H10N2O
Molecular Weight (g/mol) 102.137
MDL Number MFCD06200896
InChI Key OPENCMFJZQABIY-UHFFFAOYSA-N
Synonym n-hydroxybutyramidine, 1z-n'-hydroxybutanimidamide, n-hydroxybutyrimidamide, z-n'-hydroxybutyrimidamid e, butanimidamide, n-hydroxy, z-n'-hydroxybutimidamide, butanimidamide,n-hydroxy, e-n'-hydroxybutanimidamide, 1-1h-benzo d imidazol-2-yl-3-methyl-4-oxo-1,4-diShow MoreShow Less
PubChem CID 9582833
IUPAC Name N'-hydroxybutanimidamide
SMILES CCCC(=NO)N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 102.14
Percent Purity 97%
Chemical Name or Material N-hydroxy-butyramidine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
n-Hydroxy-butyramidine, ≥97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.