missing translation for 'onlineSavingsMsg'
Learn More
N-Carbamoylmaleamic acid, 98%, Thermo Scientific™ Product Code.: 11407034

N-Carbamoylmaleamic acid, 98%, Thermo Scientific™

Product Code. 11407034
10 g
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy

Product Code. 11407034

Brand: Thermo Scientific Alfa Aesar B21487.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
N-Carbamoylmaleamic acid, is used as a pharamceutical intermediate.

Solubility
Soluble in hot acetic acid, ethanol, acetone, benzene, chloroform. Practically insol in cold water, cold acetic acid, acetone, ligroin, chloroform, alc, ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-61-3
Molecular Formula C5H6N2O4
Molecular Weight (g/mol) 158.113
MDL Number MFCD00014790
InChI Key GWGLGTKSTGSWGQ-UPHRSURJSA-N
Synonym n-carbamoylmaleamic acid, maleuric acid, maleylurea, maleic acid monoureide, maleinuric acid, unii-j7d540jrqc, j7d540jrqc, 4-aminocarbonyl amino-4-oxo-2-butenoic acid, n-carbamoyl-maleamic acid, 2z-4-aminocarbonylamino-4-oxobut-2-enoic acidShow MoreShow Less
PubChem CID 5356398
IUPAC Name (Z)-4-(carbamoylamino)-4-oxobut-2-enoic acid
SMILES C(=CC(=O)O)C(=O)NC(=O)N

Specifications

Melting Point 156°C to 159°C
Quantity 10 g
Beilstein 1724683
Merck Index 14,5706
Solubility Information Soluble in hot acetic acid,ethanol,acetone,benzene,chloroform. Practically insol in cold water,cold acetic acid,acetone,ligroin,chloroform,alc,ether.
Formula Weight 158.11
Percent Purity 98%
Chemical Name or Material N-Carbamoylmaleamic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberE4935A Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Lot NumberP04831 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-Carbamoylmaleamic acid, 98%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.