missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Cyanoethyl)glycine, 98%, Thermo Scientific™

Product Code. 10571401 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 10571401

Brand: Thermo Scientific Acros 154900250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 3088-42-4
Molecular Formula C5H8N2O2
Molecular Weight (g/mol) 128.13
MDL Number MFCD00066903
InChI Key KZUBZCHAWPDYQX-UHFFFAOYSA-N
Synonym n-2-cyanoethyl glycine, glycine, n-2-cyanoethyl, n-2-cyanoethyl aminoacetic acid, 2-2-cyanoethyl amino acetic acid, 2-2-cyanoethylamino acetic acid, 2-cyanoethyl amino acetic acid, wln: qv1m2cn, .alpha.-.beta.'-cyanoethyl-aminoacetic acid
PubChem CID 76526
IUPAC Name 2-(2-cyanoethylamino)acetic acid
SMILES OC(=O)CNCCC#N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 188°C to 193°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula NCCH2CH2NHCH2CO2H
Quantity 25 g
Formula Weight 128.13
Percent Purity 98%
Physical Form Powder or Flakes
Chemical Name or Material N-(2-Cyanoethyl)glycine, 98%
Show More Show Less
compliance-icons
Product Identifier
  • N-(2-Cyanoethyl)glycine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.