Learn More
MYO1B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15012-1-AP-20UL
 View more versions of this product
Product Code. 16803984
  • € 145.00 / 20µL
Description
Motor protein that may participate in process critical to neuronal development and function such as cell migration, neurite outgrowth and vesicular transport.
Specifications
Specifications
MYO1B
Polyclonal
Unconjugated
MYO1B
MMI alpha, MMIa, MYH 1c, MYO1B, Myosin I alpha, myosin IB, myr1
Rabbit
Antigen Affinity Chromatography
RUO
117057, 17912, 4430
-20°C
Liquid
Immunohistochemistry (Paraffin)
0.29 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O43795, P46735, Q05096
Myo1b
MYO1B Fusion Protein Ag6895
20 μL
Primary
Human, Mouse, Rat