Learn More
Thermo Scientific™ Multidrop™ Combi Reagent Dispenser
Product Code. p-4528697 Shop All Thermo Scientific Products
Click to view available options
Description:
Multidrop Combi SMART+
Multidrop Combi w/ SMART 2 Option
Multidrop Combi+
Unit Size:
Each
Product Code. 15377400
Brand: Thermo Scientific™ 5840320
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Meet your lab's specific needs with outstanding flexibility and performance with the easy-to-use Thermo ScientificTM MultidropTM Combi Reagent Dispenser.
Meet your lab's specific needs with outstanding flexibility and performance with the easy-to-use Thermo Scientific™ Multidrop™ Combi Reagent Dispenser, designed to support the varied requirements of pharmaceutical and biotechnology laboratories. It combines the most versatile features for reagent dispensing with excellent performance for drug discovery or genomic and proteomic assays.
The Multidrop Combi Reagent Dispenser handles a wide selection of plates and volume range and provides fast dispensing and high-throughput operation.
  • Provides precise dispensing over a 0.5 to 2500μL range, ensuring reproducible assay data
  • Accommodates microplates from 6 to 1536 wells and plate heights of 5 to 50mm
  • Visual icon-based graphic display makes it easy to use and program, even without training
  • Minimal dead volume and back-flushing reduces reagent costs
  • Easy to use Thermo Scientific FILLit Software provides increased flexibility and functionality
  • Full robotic compatibility gives increased throughput
  • Uses eight-channel detachable and autoclavable dispensing cassettes that are standard across the Multidrop range
Multidrop Combi SMART:
  • Adds an advanced built-in tracing system to the Multidrop Combi
  • Provides improved realiability and cassette lifetime traceability, greatly enhancing user efficiency and reporting capability
Recommended for:
  • Assay development
  • Primary and secondary screening
  • Compound storage
  • Genomics and proteomics research
  • PCR set-up
  • Sequencing set-up
  • Cell based assays
  • Bead based assays
  • ELISA assays
TRUSTED_SUSTAINABILITY
Specifications
Accuracy Small tube dispensing cassette: 2μL, ±10% (typical); 10μL, ±5% (typical); >10μL, ±5% (typical)
Stand tube dispensing cassette: 5μL, ±3%(typical); 20μL, ±2% (typical); 100μL, ±1% (typical); >100μL, ±1% (typical)
Product Type Reagent Dispenser
Description Multidrop Combi w/ SMART 2 Option
Channels 8
Volume (Metric) 0.5 to 2500 μL
Increments Small tube dispensing cassette: 0.5μL increments 0.5-50μL
Stand tube dispensing cassette: 5μL increments 5-2500μL
Interface Serial RS-232, USB
Plate Type 6- to 1536-well plates
Precision Small tube dispensing cassette: 0.5μL, CV ≤10%; 2μL, CV≤5%; 10μL, CV≤3%; >10μL, CV≤3%
Stand tube dispensing cassette: 5μL, CV≤10%; 20μL, CV≤1.5%; 100μL, CV≤1%; >100μL, CV≤1%
Speed 96-well plate: 10μL in 3 sec.; 20μL in 4 sec.; 100μL in 10 sec
384-well plate: 1μL in 5 sec.; 5μL in 5 sec.; 10μL in 6 sec.; 20μL in 9 sec.
1536-well plate: 1μL in 14 sec.; 5μL in 26 sec.
Volume (Metric) Dead <1 mL volume range 0.5to 50 μL; <7 mL volume range >50 μL
Width (English) 14 in.
Width (Metric) 35.5 cm
Height (English) 8.6 in.
Height (Metric) 22 cm
Depth (English) 12.9 in.
Depth (Metric) 33 cm
Weight (English) 20.1 lb.
Weight (Metric) 9.1 kg
Electrical Requirements 100 to 240V 50/60Hz
Voltage 100/240 V
Frequency 50/60 Hz
Mostrar más Mostrar menos
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.