Learn More
Tocris Bioscience™ Mps1-IN-1 dihydrochloride
Selective Mps1 kinase inhibitor
Brand: Tocris Bioscience™ 5142/10
320.51 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
Mps1-IN-1 dihydrochloride is a selective monopolar spindle 1 (Mps1) kinase inhibitor (IC50 = 367 nM); exhibits >1000 fold-selectivity against a panel of 352 kinases with the exceptions of ALK and Ltk. Disrupts the recruitment of Mad2 to kinetochores. Increases frequency of multipolar mitosis in U2OS cells.Specifications
>98% | |
10 mg | |
608.58 |
1883548-93-3 | |
Mps1-IN-1 dihydrochloride | |
Mps1-IN-1 dihydrochloride |
For Research Use Only