Learn More
Tocris Bioscience™ Mpro 13b-H
Negative control for Mpro 13b-K (Cat. No. 7228)
Brand:  Tocris Bioscience™ 7853/5
241.03 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18748716
  • € 273.00 / 5mg
Explore more special offers
Description
Description
Mpro 13b-H is a negative control for the coronavirus Mpro inhibitor, Mpro 13b-K (Cat. No. 7228).
Specifications
Specifications
tert-Butyl (1-((R)-1-(((S)-4-(benzylamino)-3,4-dioxo-1-((S)-2-oxopyrrolidin-3-yl)butan-2-yl)amino)-3-cyclopropyl-1-oxopropan-2-yl)-2-oxo-1,2-dihydropyridin-3-yl)carbamate
5 mg
3C and 3CL Protease Inhibitors
C31H39N5O7