Learn More
Plum™ MP2000 Dispenser
Product Code. p-7159695
Click to view available options
Width (Metric):
130 mm
230 mm
Unit Size:
Each
Product Code. 15761137
Brand: Plum™ P4251
Only null left
Add to basket
Add to basket
Flexible dispenser system for bag-in-box products (soap/ hand cleaning/ hand lotion). Well-suited for production plants, repair shops, food processing, technical colleges and graphical industries.
  • Hygienic and reliable dispenser design
  • Easily customised to hand hygiene requirements in the individual workplace
  • MP2000 products are supplied in airtight packages for prevention of bacteria growth
  • Easily mounted, cleaned and refilled
  • Specifications
    Depth (Metric) 110 mm
    Width (Metric) 130 mm
    Height (Metric) 300 mm
    No. of Compartments 1