Learn More
MP Biomedicals™ Terrific Broth II, Dehydrated Media in Pouches
Shop All MP Biomedicals Products
Click to view available options
:
Makes 10 x 1L
Specifications
Specifications
| Certifications/Compliance | Molecular Biology Certified™ |
| Content And Storage | 15°C to 30°C |
| Form | Dehydrated |
| Grade | Molecular Biology |
| Product Type | Microbiology Media |
| Formulation | Yeast extract, Tryptone broth, Sodium Chloride and Vitamins |
| For Use With (Application) | For growth of Escherichia coli |
| Includes | 10 × 50g pouches |
| Packaging Type | Pouch |
| Quantity | 10 x 1000 mL |