missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse LYPD6 (NP_796113) VersaClone cDNA
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Description
- The Mouse LYPD6 (NP_796113) VersaClone cDNA clone contains the complete ORF for LYPD6, along with a Kozak consensus sequence for optimal translation initiation
- The gene insert is flanked with convenient multiple cloning sites, which can be used to easily cut and transfer the gene cassette into your desired expression vector
- The LYPD6 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided
Specifications
Specifications
| Gene | mLYPD6 |
| Accession Number | NP_796113 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Gene ID (Entrez) | 130574 |
| Content And Storage | Store the unopened product at -20 to -80°C |
| Species | Mouse |
| Quantity | 10 μg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction