missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech™ Mouse IL-10 ELISA Kit
KE10008 is a solid phase sandwich Enzyme Linked-Immuno-Sorbent Assay (Sandwich ELISA). The mouse IL10 ELISA kit is to be used to detect and quantify protein levels of endogenous mouse IL10.
Brand: Proteintech™ KE10008-96T
This item is not returnable.
View return policy
Description
- The assay recognizes mouse IL10. An antibody specific for mouse IL10 has been pre-coated onto the microwells.
- The mouse IL10 protein in samples is captured by the coated antibody after incubation. Following extensive washing, another antibody of biotinylated specific for mouse IL10 is added to detect the captured mouse IL10 protein.
- For signal development, Streptavidin-HRP is added, followed by Tetramethyl-benzidine (TMB) reagent.
- Solution containing sulfuric acid is used to stop color development and the color intensity which is proportional to the quantity of bound protein is measurable at 450nm with the correction wavelength set at 630 nm.
- Interleukin-10 is an anti-inflammatory cytokine, produced by T helper (Th) cells, macrophages, monocytes, and B cells, that plays a crucial role in preventing inflammatory and autoimmune pathologies.
- It down regulates the expression of Th1 cytokines, MHC class IL antigens, and co-stimulatory molecules on macrophages.
- It also enhances B cell survival, proliferation, and antibody production. IL-10 can block NF-κB activity, and is involved in the regulation of the JAK-STAT signaling pathway. IL-10, along with its receptors, describes an important role in pathogenesis of various diseases, including infectious, inflammatory, autoimmune diseases.
- IL-10 mutations are associated with an increased susceptibility to HIV-1 infection and rheumatoid arthritis.
Specifications
NP_034678 | |
2.5 pg/mL | |
Sandwich ELISA, Double Antibody | |
Plasma, Serum | |
Mouse | |
Mouse | |
<10% | |
Blue ice | |
4.5 hr. |
15.6 to 1000 pg/mL | |
ISO 13485 | |
Coated ELISA Kit | |
DAB | |
16153 | |
Solid | |
<10% | |
-20°C to +4°C | |
P18893 |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- 96TEST Human COMP ELISA Kit
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 1
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
Precautionary Statement
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P234-Keep only in original container.
- P264-Wash skin thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P330-Rinse mouth.
- P362-Take off contaminated clothing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Sulphuric Acid, Tetramethylbenzidine