missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse IgG1 Isotype Control, Pacific Orange™
Mouse Isotype Control
Brand: Invitrogen™ MG130
This item is not returnable.
View return policy
Description
Mouse IgG1 Isotype Control for Flow
Mouse IgG1 Control antibody specifically detects Mouse IgG1 in Not Applicable samples. It is validated for Flow Cytometry.Specifications
Mouse IgG1 | |
Pacific Orange | |
IgG1; Igh-4; Ighg1; Immunoglobulin G; Immunoglobulin G1; immunoglobulin heavy constant gamma 1 (G1m marker); ImmunoglobulinG; ImmunoglobulinG1; VH7183 | |
purified | |
ASR | |
Isotype Control (Primary) | |
2-8°C | |
IgG1 | |
Wet Ice |
Flow Cytometry | |
PBS with 0.1% sodium azide | |
Mouse | |
500 μL | |
Whole Antibody | |
Not Applicable | |
Liquid | |
50 Tests | |
Pacific Orange™ |