missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse GnRH1 Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP168
This item is not returnable.
View return policy
Description
Gonadotropin Releasing Hormone (GnRH) Synthetic Peptide, PEP-168, from Invitrogen.
PEP-168 is a 10 amino acid synthetic peptide whose sequence corresponds to GnRH. The sequence of this peptide is (amino to carboxy terminus): pyroE-H-W-S-Y-G-L-R-P- (G-NH2)This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and GnRH, catalog # PA1-120. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∼70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 ml of distilled water.Specifications
14714 | |
purified | |
Mouse GnRH1 | |
-20°C. Avoid freeze-thaw cycles. | |
Lyophilized | |
Control | |
Yes | |
Peptide | |
RUO - research use only |
Mouse | |
P13562 | |
0.5 mg/mL | |
Synthetic Peptide | |
peptide with no preservative | |
Gnrh1 | |
No preservative | |
50 μg | |
pyroE-H-W-S-Y-G-L-R-P-(G-NH2) |