missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse DCIR/CLEC4A (NP_036129) VersaClone cDNA
This shuttle vector contains the complete ORF for the gene of interest, along with a Kozak consensus sequence for optimal translation initiation. It is inserted NotI to AscI. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector.
Brand: R&D Systems™ RDC2581
This item is not returnable.
View return policy
Description
- Clec4a2 C-type lectin domain family 4, member a2 [ Mus musculus (house mouse) ]
- Also known as: DCIR; Dcir1; Clec4a; Clecsf6
- DCIR, also known as lectin-like immunoreceptor (LLIR), is a type II transmembrane protein belonging to the C-type lectin domain family and is designated CLEC4A (previously designated CLECSF6).
- Four transcript variants encoding distinct isoforms have been identified.
- DCIR contains one carbohydrate recognition domain in its C-terminal extracellular domain and an immunoreceptor tyrosine-based inhibitory motif (ITIM) in its cytoplasmic domain.
- DCIR is expressed on dendritic cells, B cells, monocytes/macrophages and granulocytes.
- The DCIR/CLEC4A insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
mClec4a2 | |
10 μg at 0.2 μg/μL | |
730 bp | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
10 μg |
NP_036129 | |
cDNA is provided in 10 mM Tris-Cl, pH 8.5 | |
50856 | |
Mouse |