Learn More
GIBCO™ Mouse (Balb-c) Microsomes
Liver microsomes prepared from a pool of male Balb-c Mice. Total protein concentration: 20 mg/mL.
Brand: GIBCO™ BCMCPL
Specifications
Pooled | |
Frozen | |
0.5 mL | |
20 mg/mL | |
Store between -60°C and -85°C. | |
Microsomes | |
Gibco™ |
Male | |
Liver | |
Mouse | |
Mouse Microsome | |
Tube | |
Dry Ice |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 8
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2513990-A | Certificate of Analysis | 29/01/2025 | BCMCPL |
2513990-B | Certificate of Analysis | 29/01/2025 | BCMCPL |
2912182 | Certificate of Analysis | 26/01/2025 | BCMCPL |
2029033-C | Certificate of Analysis | 17/02/2021 | BCMCPL |
2029033-B | Certificate of Analysis | 05/12/2019 | BCMCPL |
Product Content Correction
Product Title
GIBCO™ Mouse (Balb-c) Microsomes > Mouse (Balb-c) Microsomes