Learn More
Invitrogen™ Vancomycin, BODIPY™ FL Conjugate (BODIPY™ FL Vancomycin)
Product Code. 11520826
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
Product Code. 11520826
Brand: Invitrogen™ V34850
BODIPY™ FL vancomycin, which contains a single BODIPY™ dye per vancoymycin molecule, is a green-fluorescent analog of this important antibiotic, which is active against gram-positive bacteria, including entereococci.
BODIPY™ FL vancomycin, which contains a single BODIPY™ dye per vancoymycin molecule, is a green-fluorescent analog of this important antibiotic, which is active against gram-positive bacteria, including entereococci. Although its biological activity has not been reported, it may be useful for detecting vancomycin binding sites and transport, for fluorescence polarization assays, and for the study and detection of vancomycin-resistant enterococci.
To make a concentrated stock solution, dissolve in either high quality anhydrous DMSO (1 mM) or H2O (∼60 mM). Store DMSO stock solutions frozen, desiccated and protected from light. Aqueous stock solutions should be used fresh as prolonged storage may result in aggregation.
Order Info
Shipping Condition: Room temperature
TRUSTED_SUSTAINABILITY
Specifications
Quantity 100 μg
Shipping Condition Room Temperature
Product Line BODIPY
Label Type BODIPY Dyes
Molecular Formula C80H88BF2Cl2N11O25
Physical Form Solid
Product Type Antibiotic
Excitation/Emission 504/511 nm
Molecular Weight (g/mol) 1723.35