missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TC-ReAsH™ II In-Cell Tetracysteine Tag Detection Kit (Red Fluorescence), for live-cell imaging

Contains an expression tag-based fluorescence labeling reagent for live-cell labeling

Brand:  Invitrogen™ T34562

Additional Details : Weight : 0.00450kg

Product Code. 10503913

  • € 1220.00 / Each
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Includes: Kit contains ReAsH-EDT2 labeling reagent and BAL wash buffer.

Description

Description

Mammalian cell lines expressing a protein fused to a tetracysteine tag (CCPGCC) can be labeled with the red-fluorescent ReAsH-EDT2 reagent contained in the kit. The tagged protein is fluorescent only when the labeling reagent is added. BAL wash buffer replaces the previously supplied Disperse Blue 3 and EDT-based wash buffer as an olfactorily more agreeable reagent that yields superior signal to noise.

Cell Analysis, Cell-Based Reporter Assays, Cellular Imaging, Enzyme and Protein Activity Assays, Labeling Chemistry, Lumio Detection, Proteins, Expression, Isolation and Analysis, Reporter Assays, Tetracysteine (Lumio/FlAsH/ReAsH) Assays

Order Info

Shipping Condition: Wet Ice

Specifications

Specifications

Tetracysteine Tag Detection Kit
Fluorescence Microscope
Red
Wet Ice
TC-ReAsH II
Fluorescent
Store in freezer (-5 to -30°C).
ReAsH
Kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.