Learn More
Invitrogen™ β-Py-C10-HPC (1-Hexadecanoyl-2-(1-Pyrenedecanoyl)-sn-Glycero-3-Phosphocholine)
Phospholipid analog of phosphocholineβ-py-C10-HPC
Brand:  Invitrogen™ H361
Product Code. 10730194
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Labeled with pyrene on the sn-2 acyl chain. Pyrene forms excimers at high concentrations, excimer emission ∽470nm. Pyrene is environment sensitive, essentially nonfluorescent in water.
Cell Analysis, Cell Metabolism, Cell Viability, Proliferation & Function, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining & Detection
Order Info
Shipping Condition: Room Temperature
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Phospholipids
1 mg
Invitrogen
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1964854 Certificate TypeCertificate of Analysis Date02/02/2018 Catalog NumberH361
Lot Number1917913 Certificate TypeCertificate of Analysis Date29/08/2017 Catalog NumberH361
Special Offers
Special Offers
Product Content Correction
Product Title
Invitrogen™ β-Py-C10-HPC (1-Hexadecanoyl-2-(1-Pyrenedecanoyl)-sn-Glycero-3-Phosphocholine) > 1mg