missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Lipopolysaccharides from Salmonella Minnesota, Alexa Fluor™ 488 conjugate
Click to view available options
Quantity:
100 μg
Unit Size:
Each
Description
Cell Analysis, Cell Cycle, Cell Structure, Cell Viability, Proliferation and Function, Endocytosis, Exocytosis and Phagocytosis, Lysosomes, Vacuoles
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Content And Storage | Store in freezer (-5 to -30°C) and protect from light. |
| Detection Method | Fluorescence |
| Product Type | Lipopolysaccharide Conjugate |
| Dye Type | Alexa Fluor Dyes |
| Product Line | Alexa Fluor |
| Quantity | 100 μg |
| Shipping Condition | Room Temperature |
| Excitation/Emission | 495/519 |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction