Learn More
OHAUS™ Modular Blocks and Accessories
Product Code. p-7078394 Shop All Ohaus Products
Click to view available options
Sample Type:
1.5mL Tube
10mm Tube
15/16mm Tube
15mL Tube
50mL Tube
Type:
Dry Block
Temperature Probe
Depth (Metric) Well:
3.91 cm
4.45 cm
4.76 cm
4.8 cm
Diameter (Metric) Well:
1.07 cm
1.1 cm
1.71 cm
1.75 cm
2.9 cm
Unit Size:
Each
To avoid possible electrical hazard, DO NOT fill well or block with water or other liquids. Units are designed as a dry bath/incubator.
Product Code. 15712579
Brand: OHAUS™ 30400159
Only null left
Add to basket
Add to basket
To avoid possible electrical hazard, DO NOT fill well or block with water or other liquids. Units are designed as a dry bath/incubator.
Constructed from solid anodized aluminum block.
Modular blocks are sold separately from heaters.
  • Close contact of tubes-to-block-walls allow for maximum heat retention
  • Each block has a thermometer well for measuring block temperature
TRUSTED_SUSTAINABILITY
Specifications
Description Module Block 1.5 mL Microtaper
For Use With (Equipment) Dry Block Heater
Material Anodized Aluminum
No. of Places 20
Type Dry Block
Sample Type 1.5mL Tube
Length (English) 3.75 in.
Length (Metric) 9.5 cm
Height (English) 2 in.
Height (Metric) 5.1 cm
Depth (English) Well 1.53 in.
Depth (Metric) Well 3.91 cm
Diameter (English) Well 0.43 in.
Diameter (Metric) Well 1.1 cm