Learn More
Miele Professional™ Mobile unit
Shop All Miele Products
Click to view available options
:
Dimensions (D x W x H): 790 x 640 x 524mm
Description
- Loading capacity: 9 DIN mesh trays
- Equipped with 2 integrated spray arms
- The loading areas for the individual levels are different
- Magnetic strip for mobile unit recognition (incl. magnets)
Specifications
Specifications
| Material | Stainless Steel |
| Product Type | Mobile Unit for Mesh Trays |
| Color | Stainless Steel |
| Holds | 9 DIN mesh trays6 SPRI mesh trays |
| For Use With (Equipment) | Washer-disinfectors, laboratory |
| Depth (Metric) | 790 mm |
| Height (Metric) | 524 mm |
| Width (Metric) | 640 mm |