Learn More
Tuttnauer Mobile Lifting Device
Mobile lifting device for vertical autoclaves with heavy loads up to 100 kg. The lift is mobile, equipped with an integrated swivel arm for maximum maneuverability and can service a few autoclaves. Controlled by push buttons on a movable handset. Equipped with a maintenance free electrical battery.
Brand:  Tuttnauer HLD507-0044
Product Code. 17829548
  • € 11140.00 / Each
Estimated Shipment: 08-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • The mobile lifting device for vertical autoclave basket with heavy sterilization load up to 100 kg.
  • The lift is mobile equipped with an integrated swivel arm for maximum maneuverability and can service a few autoclaves.
  • Controlled by push buttons on a movable control handset.
  • Equipped with a maintenance free electrical battery.
Specifications
Specifications
Mobile Lift
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Tuttnauer Mobile Lifting Device > Mobile Lifting Device