Learn More
MMAF-d8 hydrochloride, MedChemExpress
MedChemExpress D8-MMAF hydrochloride is a deuterated form of MMAF hydrochloride, which is a microtubule disrupting agent.
Brand: Medchem Express HY-15579AS
This item is not returnable.
View return policy
Description
D8-MMAF hydrochloride is a deuterated form of MMAF hydrochloride, which is a microtubule disrupting agent.Specifications
MMAF-d8 hydrochloride | |
Off-White | |
C39H58D8ClN5O8 | |
CC[C@H](C)[C@@H]([C@@H](CC(N1[C@@H](CCC1)[C@@H]([C@@H](C)C(N[C@H](C(O)=O)CC2=CC=CC=C2)=O)OC)=O)OC)N(C([C@@](C([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])([2H])NC([C@H](C(C)C)NC)=O)=O)C.Cl | |
776.47 | |
Research | |
Solid |
5 mg | |
Research | |
DMSO : 100 mg/mL (128.79 mM; Need ultrasonic) | |
776.47 | |
98.97% | |
Cancer-programmed cell death |
Research purposes only