Learn More
MMAE-d8, MedChemExpress
MedChemExpress MMAE-d8 (Monomethyl auristatin E-d8) is a deuterated labeled MMAE, a potent mitotic inhibitor and a tubulin inhibitor.
Brand: Medchem Express HY-15162A
This item is not returnable.
View return policy
Description
MMAE-d8 (Monomethyl auristatin E-d8) is a deuterated labeled MMAE, a potent mitotic inhibitor and a tubulin inhibitor.Specifications
MMAE-d8 | |
5 mg | |
Research | |
Monomethyl auristatin E-d8 Deuterated labeled MMAE | |
CC[C@H](C)[C@@H]([C@@H](CC(N1[C@@H](CCC1)[C@@H]([C@@H](C)C(N[C@H](C)[C@@H](O)C2=CC=CC=C2)=O)OC)=O)OC)N(C([C@@](C(C([2H])([2H])[2H])(C([2H])([2H])[2H])[2H])([2H])NC([C@H](C(C)C)NC)=O)=O)C | |
726.03 | |
Research | |
Solid |
2070009-72-0 | |
White | |
C39H59D8N5O7 | |
DMSO : 100 mg/mL (137.74 mM; Need ultrasonic) | |
726.03 | |
98.43% | |
Cancer-programmed cell death |
Research purposes only