missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ ML 382

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
ML 382 is a selective MRGPRX1 positive allosteric modulator (EC50 = 190 nM). Exhibits selectivity for MrgX1 over MrgX2, MrgC11 and a panel of 68 other GPCRs, ion channels and transporters. Enhances BAM (8-22) potency at MRGPRX1 by 7-fold.
Specifications
Specifications
| Chemical Name or Material | Mas-Related G Protein Coupled Receptor Modulators |
| CAS | 1646499-97-9 |
| Quantity | 10 mg |
| Target | Sensory Neuron-Specific Receptor Agonists |
| Molecular Formula | C18H20N2O4S |
| Formula Weight | 360.43 |
| Purity | >98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction