Learn More
Tocris Bioscience™ ML 337
Selective negative allosteric modulator of mGlu3
Brand: Tocris Bioscience™ 4982/50
565.00 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
ML 337 is a selective negative allosteric modulator of mGlu3 (IC50 = 593 nM). Displays no activity at mGlu1, mGlu2 or mGlu4-8 at concentrations up to 30 μM. Brain penetrant.Specifications
>98% | |
ML 337 | |
ML 337 |
50 mg | |
353.39 |
For Research Use Only