Learn More
Cytiva Mixing Chamber For use with Mixer M-925
Product Code. 12594136 Shop All Cytiva Products
Click to view available options
Volume (Metric):
2 mL
Unit Size:
Each
Product Code. 12594136
Brand: Cytiva 18111891
Only null left
Specifications
Connection Type Finger tight connector
Max. Pressure 250 bar
Volume (Metric) 2 mL
pH 1 to 13 Long Term/1 to 14 Short Term