Learn More
Gilson™ Speed Control Module
Integrates easily into existing systems with direct control through the keypad or remotely through another device. The Speed Control Module is quiet and user friendly. For use with MINIPULS™ 3 Peristaltic Pump.
Brand: Gilson™ F155001
2631.08 EUR valid until 2025-04-13
Use promo code "24122" to get your promotional price.
This item is not returnable.
View return policy
Description
- Quiet, user friendly and easy to integrate into your existing system with direct control through the keypad or remotely through another device.
- For use with MINIPULS™ 3 Peristaltic Pump.
- Flow Rate: 0.3 μL to 220 mL/min.
Specifications
MINIPULS™ 3 | |
High-torque Stepper Motor | |
Microprocessor | |
Module |
Product Content Correction
Product Title
Gilson™ Speed Control Module > Speed Control Module; Flow Rate: 0.3 μL to 220 mL/min.