Learn More
Lovibond™ Water Testing Minikit
Product Code. 10228330
Click to view available options
Range:
0 to 500 mg/L
0.75 to 10% acid
10 to 500 mg/L
2 to 50 mg/L
20 to 200 mg/L
20 to 500 mg/L
5 to 5000 mg/L
70 to 1500 mg/L
Analytes:
Alkalinity Total M
Alkalinity-P, CaC03
Chloride
Cleaning Acid Strength
Cyanuric Acid
Nitrite
Sulphate (low range) Na2SO4
Sulphite (high range)
Sulphite (low range)
Unit Size:
Each
Product Code. 10228330
Brand: Lovibond™ 414140
Lovibond™ Water Testing Minikit is easy to use and provides highly accurate results.
  • Easy operation and exact reagent dosing
  • High accuracy
  • Foil-wrapped Lovibond™ tablet reagents with a minimum guaranteed shelf life of 5 years
  • Unrestricted shipment
  • Safe storage
Specifications
Analytes Alkalinity-P, CaC03
Includes Flip top case containing a shaker tube, rapid dissolving tablet reagents and instructions.
Range 20 to 500 mg/L
Sufficient For Approximately 40 Tests
Test Method Tablet count
Type Minikit water testing kit
compliance-icons
Product Identifier
  • MINIKIT TEST ALKALINITY P AF414 .
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : sulphanilic acid