Learn More
Ekastu Safety™ Eye Wash Bottle with Funnel
Made for immediate rinsing of the eyes after accidental splashing from acids, bases and other hazardous chemicals. EKASTU Safety™ Eye Wash Bottle with Funnel can be used in either upright or horizontal positions.
€ 37.00
Specifications
Capacity (Metric) 600 mL
Includes Screwed-on Eye Funnel and Tilting Cap
Product Type Eyewash Bottle with Screwed on Funnel, Tilting Valve, Riser Pipe and Water Drain Hose
View More Specs
Products 1
Product Code Brand For Use With (Application) Price Quantity & Availability  
Product Code Brand For Use With (Application) Price Quantity & Availability  
10470903
View Documents
Ekastu Safety™
177959
Saline Solution for Immediate Rinsing of the Eyes after Contact with Acids, Bases
€ 37.00
Each
Estimated Shipment: 14-01-2025
to see stock.
Add to basket
Description
Description
  • Eye Wash Bottle with funnel
  • DIN EN 15154-4
  • Supplied empty or filled
  • For immediate rinsing of the eyes after corrosion by acids, bases and other harsh chemicals
  • PE bottle with screwed-on eye funnel and tilting valve
  • Can be used in both upright and horizontal positions
  • leak-proof
  • Stores easily in EPS Eye Wash Station (sold separately)
Specifications
Specifications
600 mL
Screwed-on Eye Funnel and Tilting Cap
Eyewash Bottle with Screwed on Funnel, Tilting Valve, Riser Pipe and Water Drain Hose