Learn More
B Braun™ Mini Spike™ V Dispensing Pin
B Braun™ Mini Spike™ Dispensing Pin can be used to withdraw or inject fluids from medication vials or semi-rigid containers.
Brand:  B Braun™ 4550560
Product Code. 15309803
  • € 4.00 / Each
Edge
Explore more special offers
Description
Description
  • Includes 0.45μm bacteria retentive air filter to reduce contamination
  • Integrated air channel enables automation of filter pressure balancing technique steps, which singnificantly increases efficiency and safety
  • Designed with a snap cap to reduce touch contamination by allowing user to work away from the critical luer connection
  • Molded tight-sealing cap improves working hygiene by preventing cap loss and potential leakage when closed
  • PVC-, DEHP, and latex-free
  • Needle-free solution to withdraw or injection fluids
  • High flow rate
  • Ergonomic design
  • 2 distinct air and fluid channels
  • Universal spike for medication vial
  • Large grip plate for convenient and intuitive handling
  • Integrated two way valve that protects against free-flow when the system is turned upside down
Specifications
Specifications
Yes
Mini Spike Dispensing Pin with Valve
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers