Learn More
Buerkle™ MiniSampler Environmental Sampling Kit
Choose from the PE model for general sampling or PTFE model for ultra pure sampling. Buerkle™ MiniSampler Environmental Sampling Kits are ideal for sampling from barrels, canisters, tanks and other large volume liquid storage units.
Brand:  Buerkle™ 5305-0100
 View more versions of this product
Product Code. 10268460
  • € 285.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Alert:
Not for aggressive acids. Vapours could destroy pump.
Description
Description
  • Sample liquid only comes into contact with the suction hose and original sample bottle minimizing possibility of cross-contamination
  • Bottles are sealed immediately after taking sample
  • Disposable suction hose can quickly be replaced ensuring samples are not contaminated
  • If necessary, a new hose can be used for each sample
  • 8 mm hose can reach narrow, seemingly inaccessible sampling areas
  • Also suitable for viscous oils
  • Complete with a handy transport case
Specifications
Specifications
Liquid Sampler
Maximum suction height: 5 m
General Sampling
Polyethylene
Vacuum pump, 100 mL 10 PE bottles, 10 m PE suction hose (6 x 8 mm), 1 hose cutter, 1 Stainless Steel hose weight made of V2A (1.4301), 20 seal-it bottle seals
Videos
  • Buerkle MiniSampler Environmental Sampling Kit
    Buerkle MiniSampler Environmental Sampling Kit
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Buerkle MiniSampler Environmental Sampling Kit
SDS
Documents
Documents
Brochures
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Buerkle™ MiniSampler Environmental Sampling Kit > 100mL Vacuum Pump; 10 PE bottles; 10m PE suction hose (6 x 8mm)