Learn More
Brady™ Mild Sheet Steel Portable Metal Group Lock Box
Product Code. p-8001140
Click to view available options
Color:
Red
Yellow
Unit Size:
Each
Product Code. 11751915
Brand: Brady™ 065672
Only null left
Add to basket
Add to basket
Simplifies lockout of large equipment
  • Heavy steel lock box stores keys after machine or process is locked out
  • Each authorized work team member adds a personal lock or tagout device on the group lock box
  • Ensures that no one employee has access unless all employees have removed their locks or tags
  • Accepts up to 13 individual locks
Specifications
Product Type Portable Metal Group Lock Box
Alternate Part Number STEEL PORTABLE GROUP LOCK BOX YLW
Color Yellow
Quantity 1 Ea.