Learn More
Labcon™ Microtube Floating Storage Racks
Click to view available options
Size:
24-place
48-place
Closure Included:
Clear Lid
None
Unit Size:
Pack of 10
Pack of 5
Description
- Holds 1.5, 1.7, or 2 mL microtubes
- Freezable to -80°C
- Made in USA
Specifications
Specifications
| Closure Included | Clear Lid |
| Autoclavable | Autoclavable |
| Color | White |
| Holds | 1.5, 1.7, 2 mL Microtubes |
| Material | Polypropylene |
| Size | 48-place |