Learn More
Berkshire™ MicroSeal Supersorb™ Flat Mop Wipers
MicroSeal SuperSorb™ is a sealed edge cleanroom laundered 100% polyester knit wiper.
€ 1190.00
Specifications
Product Type | Flat Head Mop Cover |
---|---|
Sterility | Non-sterile |
Autoclavable | Autoclavable |
Description
- Critically low particles, fibers, ions and extractables
- High abrasion resistance
- Chemically compatible with IPA, Acetone and other solvents
- Highest sorbency level available in a polyester knit reduces wiper usage
- Pinsonic design adds surface texture that aids in stubborn residue removal
- Low cost in use compared to other two-ply polyester knits
Specifications
Flat Head Mop Cover | |
Non-sterile | |
Autoclavable |