Learn More
Carl Zeiss™ AxioCams Microscopy Camera
Product Code. p-8006878
Click to view available options
Description:
AxioCam 105 Color
Unit Size:
Each
Product Code. 15295639
Brand: Carl Zeiss™ 4265550000000
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
5 megapixel color microscope camera for documentation in routine labs
  • CMOS microscope camera, 5 megapixels (2560 x 1920)
  • Sensor size of 1/2.5 in. (diagonal 7.1 mm)
  • Pixel size 2.2 x 2.2 μm
  • Color camera
  • Includes ZEN Lite starter software for image capture and making measurements
Specifications
Type Microscopy Camera AxioCam
Description AxioCam 105 Color
Includes Axiocam 105 color, driver software 64-bit, USB 3.0 PCIe x1 interface, USB 3.0 connection cable (3 m), SATA power cable and Molex power cable sensor