Learn More
Thermo Scientific™ Microplate Rack Inserts for MaxQ™ 8000 Shakers
Maximize efficiency in the lab by using Thermo Scientific™ Microplate Rack Inserts for MaxQ™ 8000 shakers, which allow shaking of multiple microwell plates at one time.
Brand: Thermo Scientific™ 600089
This item is not returnable.
View return policy
Description
Maximize efficiency in the lab by using Thermo Scientific™ Microplate Rack Inserts for MaxQ™ 8000 Shakers, which allow the shaking of multiple microwell plates at one time. With ultimate flexibility, these microplate rack inserts are the optimal choice for high throughput labs.
Specifications
Microplate rack inserts, deep-well plates | |
Microplate rack inserts, deep-well plates | |
Deep-well plates | |
5 Racks, 30 plates |