Learn More
Thermo Scientific™ Microplate Rack Inserts for MaxQ™ 8000 Shakers
Maximize efficiency in the lab by using Thermo Scientific™ Microplate Rack Inserts for MaxQ™ 8000 shakers, which allow shaking of multiple microwell plates at one time.
Brand:  Thermo Scientific™ 600090
 View more versions of this product
Product Code. 10175752
  • € 396.98 / Each
Estimated Shipment: 21-05-2025
to see stock.
Explore more special offers
Description
Maximize efficiency in the lab by using Thermo Scientific™ Microplate Rack Inserts for MaxQ™ 8000 Shakers, which allow the shaking of multiple microwell plates at one time. With ultimate flexibility, these microplate rack inserts are the optimal choice for high throughput labs.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Microplate rack inserts, standard plates
Microplate rack inserts, standard plates
Standard plates
7 Racks, 63 plates