Learn More
Micronova™ Irradiated MegaSwat™ Mop Covers
GREENER_CHOICE
Product Code. p-3142926 Shop All Micronova Products
Click to view available options
Material:
Polyester
Smooth Polyester, Urethane Foam
Dimensions:
7.6 x 20.3 cm (3 x 8 in.)
7.6 x 40.6 cm (3 x 16 in.)
For Use With (Equipment):
QDMS-16 and SSU Series Handle
Unit Size:
Pack of 12
Product Code. 13177436
Brand: Micronova™ MS68IR
Only null left
Add to basket
Add to basket
Micronova™'s irradiated, mop covers work with the blade style MegaSwat™ Mop adapter to effectively clean around, under and behind heavy processing equipment and calibrated apparatus in the lab and cleanroom.
The Micronova™ Irradiated MegaSwat™ Mop Covers are designed to work with the MegaSwat™ Mop Adapter and SSU series QD handles, for wet or dry cleaning and disinfecting in ISO 5 to 9 areas. The flexible blade style mop is ideal for cleaning under, around and behind heavy or calibrated equipment in the cleanroom. The covers are gamma irradiated and available with or without a foam interior.
  • Knit polyester mops easily glide under and behind equipment
  • Designed for use with flexible MegaSwat™ Mop Adapter and SSU series QD handles
  • Available in various lengths
  • Available with or without foam interiors
  • NovaPoly, 100% polyester knit provides excellent disinfectant dispersal
  • Compatible with alcohol and strong disinfectants
  • Launderable for reuse
  • Autoclavable
Specifications
Dimensions 7.6 x 20.3 cm (3 x 8 in.)
Product Type Mop Cover
Color White
Length (English) 8 in.
Sterility Sterile
Length (Metric) 20.3 cm
Autoclavable Autoclavable
Width (Metric) 7.6 cm
Width (English) 3 in.
Disposable Reusable
Quantity Packaged in Class 100 Cleanroom. Double Bagged. Irradiation Indicator Placed on Outer Bag. 12 Mops are then Packed into a Poly Lined Case. Irradiation Indicator Placed on Case.
Care Instructions Item may be Laundered and Reused. Care should be taken to fully eliminate disinfectants prior to reuse or steam sterilization. Water or Dryer temperature should not exceed 180°C.
For Use With (Equipment) QDMS-16 and SSU Series Handle
For Use With (Application) Cleaning and Sanitizing in and around Process and Lab Equipment in the Aspetic area
Color-coded No
Material Polyester