Learn More
Hettich Lab Technology™ Microlitre Centrifuge
The Microlitre Centrifuge can process very small samples at very high speeds. The powerful, maintenance free motors achieve high RCF values for best sample results. Reliable and steady temperature control, quick pre-cool and stand-by cooling functions maintain a constant sample temperature.
Brand:  Hettich Lab Technology™ 2405
 View more versions of this product
Product Code. 10274042
  • € 5635.00 / Each
Explore more special offers
Description
Description
  • Better results and more samples in less time - increase efficiency and throughput in your lab
  • Wide range of capabilities due to the variety of models and accessories available
  • Pre-cooling function for fast operation - reliable and steady temperature control systems with quick pre-cool and standby functions for precise temperature control
Specifications
Specifications
30 x 1.5/2 mL
21,382 x g
Microlitre Centrifuge
547 mm
MIKRO 200R
15,000 rpm
-10°C to +40°C
Lid dropping protection
Yes
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Special Offers
Special Offers