Learn More
Ansell™ Microchem™ 4000 Coveralls with Hood and Face Seal
Product Code. 15526464 Shop All Ansell Products
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 8
Product Code. 15526464
Brand: Ansell™ GR40T0015107G02
Only null left
Add to basket
Add to basket
Fits full-face respirators and creates a close seal. Ansell Microchem™ 4000 Coveralls with Hood and Face Seal protect against dangerously concentrated chemicals and biological agents.
  • Coverall with elasticated 2-piece hood with neoprene rubber face seal
  • Attached Ansell Barrier Gloves, with oversleeves and finger loops
  • Integrated socks and boot overflap
  • Rear horizontal zip entry
  • Elasticated double cuffs (knitted inner cuff), waist and ankles.
  • Permeation tested against over 180 chemicals, including chemical warfare agents
  • Textile-like inner improves wearer acceptance
  • Anti-static according to EN 1149-5
TRUSTED_SUSTAINABILITY
Specifications
Product Type Protective Coveralls
Size 3X-Large
Gender Unisex
Material Multi-layer Barrier Laminate Fabric
Color Green
Disposable Limited-use
Waist Style Elastic
Ankle Style Elastic, Attached Socks, Boot Overflaps
Cuff Style Double, Elastic, Attached Gloves
Neck Style Hooded
Hood Type Attached, Neoprene Rubber Face Seal
Closure Type Rear-entry Horizontal Zipper
Seam Type Taped, Welded
Sterility Non-sterile
Resistance Chemicals
Certifications/Compliance Type 3/4 EN14605, Type 5 EN ISO 13982-1, EN14126, EN1073-2, EN1149-5
Description Hood with neoprene face seal, attached Ansell Barrier™ Gloves, integrated socks with boot overflap
No. of Pockets None
Style Includes Hood with neoprene face seal, attached Ansell Barrier™ Gloves, integrated socks with boot overflap