Learn More
Ansell™ Microchem™ 4000 Coveralls with Hood and Face Seal
Shop All Ansell Products
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 8
Description
- Coverall with elasticated 2-piece hood with neoprene rubber face seal
- Attached Ansell Barrier Gloves, with oversleeves and finger loops
- Integrated socks and boot overflap
- Rear horizontal zip entry
- Elasticated double cuffs (knitted inner cuff), waist and ankles.
- Permeation tested against over 180 chemicals, including chemical warfare agents
- Textile-like inner improves wearer acceptance
- Anti-static according to EN 1149-5
Specifications
Specifications
| Product Type | Protective Coveralls |
| Size | 2X-Large |
| Gender | Unisex |
| Material | Multi-layer Barrier Laminate Fabric |
| Color | Green |
| Disposable | Limited-use |
| Waist Style | Elastic |
| Ankle Style | Elastic, Attached Socks, Boot Overflaps |
| Cuff Style | Double, Elastic, Attached Gloves |
| Neck Style | Hooded |
| Show More |